2,5-Dichloro-4,6-dimethylnicotinonitrile 98%

Supplier: Ambeed
Warning

A153281-100G A153281-1G A153281-25G A153281-500G A153281-5G
77247-358EA 99.91 USD
77247-358 77247-360 77247-362 77247-364 77247-366
2,5-Dichloro-4,6-dimethylnicotinonitrile 98%
2,5-Dichloro-4,6-dimethylnicotinonitrile

Precaución: for Research Use Only

Formula: C₈H₆Cl₂N₂
MW: 201.0500 g/mol
MDL Number: MFCD00052631
CAS Number: 91591-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR