2-Chloro-1-fluoro-4-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-fluoronitrobenzene, 1-Chloro-2-fluoro-5-nitrobenzene

A153239-100G A153239-25G A153239-500G
77286-192EA 13.39 USD
77286-192 77286-194 77286-196
2-Chloro-1-fluoro-4-nitrobenzene 98%
2-Chloro-1-fluoro-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
MDL Number: MFCD00007206
CAS Number: 350-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR