3,4-Dimethylbenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Hydroxymethyl)-o-xylene, (3,4-Dimethylphenyl)methanol

A153236-10G A153236-1G A153236-250MG A153236-25G A153236-5G
77273-036EA 41.2 USD
77273-036 77273-038 77273-040 77273-042 77273-044
3,4-Dimethylbenzyl alcohol 98%
3,4-Dimethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₂O
MW: 136.19 g/mol
MDL Number: MFCD00004647
CAS Number: 6966-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR