2,3,4,6-Tetra-O-benzyl-α-D-glucopyranose 97%

Supplier: Ambeed
Warning

Synonyms: (2S,3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol

A152905-10G A152905-25G
77247-338EA 8.24 USD
77247-338 77247-340
2,3,4,6-Tetra-O-benzyl-α-D-glucopyranose 97%
2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranose

Precaución: for Research Use Only

Formula: C₃₄H₃₆O₆
MW: 540.65 g/mol
MDL Number: MFCD00023849
CAS Number: 6564-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR