4-Bromo-m-toluidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-methylaniline hydrochloride

A152903-100MG A152903-1G A152903-250MG A152903-5G
77286-184EA 23.69 USD
77286-184 77286-186 77286-188 77286-190
4-Bromo-m-toluidine hydrochloride 97%
4-Bromo-m-toluidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉BrClN
MW: 222.51 g/mol
MDL Number: MFCD00070748
CAS Number: 202925-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR