Ir(p-CF3-ppy)3 97%

Supplier: Ambeed
Warning

A152900-100MG A152900-250MG A152900-50MG
77247-334EA 139 USD
77247-334 77247-336 77746-052
Ir(p-CF3-ppy)3 97%
Ir(p-CF3-ppy)3

Precaución: for Research Use Only

Formula: C₃₆H₂₁F₉IrN₃
MDL Number: MFCD31657272
CAS Number: 500295-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR