4-Iodo-2-methoxy-5-methylpyridine 97%

Supplier: Ambeed
Warning

A152710-100MG A152710-250MG
77235-496EA 103 USD
77235-496 77235-498
4-Iodo-2-methoxy-5-methylpyridine 97%
4-Iodo-2-methoxy-5-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₈INO
MW: 249.04 g/mol
MDL Number: MFCD16611483
CAS Number: 1227602-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR