2-Amino-1H-benzo[d]imidazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A152663-100MG A152663-250MG A152663-1G
77247-328EA 67.98 USD
77247-328 77247-330 77746-051
2-Amino-1H-benzo[d]imidazole-5-carboxylic acid 97%
2-Amino-1H-benzo[d]imidazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD03840776
CAS Number: 76391-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR