3-((5-(2,3-Dichlorophenyl)-1H-tetrazol-1-yl)methyl)pyridine hydrochloride 98%
Supplier: Ambeed
|
Danger
|
A152651-10MG
A152651-1MG
A152651-50MG
A152651-5MG
77235-488EA
96
USD
77235-488
77235-490
77235-492
77235-494
3-((5-(2,3-Dichlorophenyl)-1H-tetrazol-1-yl)methyl)pyridine hydrochloride 98%
3-((5-(2,3-Dichlorophenyl)-1H-tetrazol-1-yl)methyl)pyridine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₃H₁₀Cl₃N₅ |
MDL Number:
MFCD09971142 CAS Number: 899431-18-6 UN: 1325 ADR: 4.1,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...