3-Nitro-1H-pyrrolo[3,2-c]pyridine 95%
Supplier: Ambeed
|
Warning
|
A152483-100MG
A152483-1G
77286-180EA
54.59
USD
77286-180
77286-182
3-Nitro-1H-pyrrolo[3,2-c]pyridine 95%
3-Nitro-1H-pyrrolo[3,2-c]pyridine
Precaución: for Research Use Only
Formula:
C₇H₅N₃O₂ |
MDL Number:
MFCD08690125 CAS Number: 23612-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...