(5aR,10bS)-2-Mesityl-4,5a,6,10b-tetrahydro-2H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-11-ium tetrafluoroborate ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A152435-100MG
A152435-250MG
77247-316EA
66.95
USD
77247-316
77247-318
(5aR,10bS)-2-Mesityl-4,5a,6,10b-tetrahydro-2H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-11-ium tetrafluoroborate ≥97%, ee 99%
(5aR,10bS)-2-Mesityl-4,5a,6,10b-tetrahydro-2H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-11-ium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₂₁H₂₂BF₄N₃O MW: 419.22 g/mol |
MDL Number:
MFCD30489132 CAS Number: 925706-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...