Sodium 2-((4-formylphenyl)(methyl)amino)ethanesulfonate 70%

Supplier: Ambeed
Warning

A152408-100G A152408-10G A152408-5G A152408-25G
77259-286EA 18.54 USD
77259-286 77259-288 77259-290 77567-202
Sodium 2-((4-formylphenyl)(methyl)amino)ethanesulfonate 70%
Sodium 2-((4-formylphenyl)(methyl)amino)ethanesulfonate

Precaución: For research use only.

Formula: C₁₀H₁₂NNaO₄S
MW: 265.26 g/mol
MDL Number: MFCD30729995
CAS Number: 56405-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR