4-(4-Fluoro-3-((pyridin-2-ylmethyl)carbamoyl)phenyl)-N-(1H-indazol-5-yl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
CCG215022
A152319-10MG
A152319-1MG
A152319-25MG
A152319-50MG
A152319-5MG
77259-270EA
375.95
USD
77259-270
77259-272
77259-274
77259-276
77259-278
4-(4-Fluoro-3-((pyridin-2-ylmethyl)carbamoyl)phenyl)-N-(1H-indazol-5-yl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxamide 98%
4-(4-Fluoro-3-((pyridin-2-ylmethyl)carbamoyl)phenyl)-N-(1H-indazol-5-yl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxamide
Precaución: for Research Use Only
Formula:
C₂₆H₂₂FN₇O₃ MW: 499.5 g/mol |
MDL Number:
MFCD30489722 CAS Number: 1813527-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...