4-Bromo-2-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-fluorobenzeneboronic acid

A152093-100MG A152093-10G A152093-1G A152093-250MG A152093-5G
77286-166EA 10.3 USD
77286-166 77286-168 77286-170 77286-172 77286-174
4-Bromo-2-fluorophenylboronic acid 98%
4-Bromo-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
MW: 218.82 g/mol
MDL Number: MFCD01318110
CAS Number: 216393-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR