5-Chloropicolinonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2-cyanopyridine

A152038-1G A152038-5G
77273-014EA 6.18 USD
77273-014 77273-016
5-Chloropicolinonitrile 98%
5-Chloropicolinonitrile

Precaución: for Research Use Only

Formula: C6H3ClN2
MW: 138.55 g/mol
MDL Number: MFCD03788835
CAS Number: 89809-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR