2-(o-tolyl)pyridine 95%

Supplier: Ambeed
Warning

A151970-1G A151970-250MG A151970-5G
77247-308EA 78 USD
77247-308 77247-310 77247-312
2-(o-tolyl)pyridine 95%
2-(o-tolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁N
MDL Number: MFCD06801884
CAS Number: 10273-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR