Fmoc-D-Tic-OH 97%

Supplier: Ambeed
Warning

A151929-100G A151929-10G A151929-1G
77286-134EA 219 USD
77286-134 77286-136 77286-138
Fmoc-D-Tic-OH 97%
Fmoc-D-Tic-OH

Precaución: for Research Use Only

Formula: C₂₅H₂₁NO₄
MDL Number: MFCD00144345
CAS Number: 130309-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR