5-Fluoro-2-(trifluoromethyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

A151728-1G A151728-250MG A151728-5G
77273-008EA 22.66 USD
77273-008 77273-010 77273-012
5-Fluoro-2-(trifluoromethyl)phenylboronic acid 97%
5-Fluoro-2-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₄O₂
MW: 207.9180 g/mol
MDL Number: MFCD06656275
CAS Number: 928053-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR