Nα-Fmoc-Nω-(p-toluenesulfonyl)-L-arginine 97%

Supplier: Ambeed
Warning

Synonyms: Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-tosyl-L-arginine, Nalpha-Fmoc-Nomega-tosyl-L-arginine, Nα-Fmoc-Nω-tosyl-L-arginine, Fmoc-Arg(Tos)-OH

A151713-100G A151713-1G A151713-25G A151713-5G
77286-122EA 247.2 USD
77286-122 77286-124 77286-126 77286-128
Nα-Fmoc-Nω-(p-toluenesulfonyl)-L-arginine 97%
Nα-Fmoc-Nω-(p-toluenesulfonyl)-L-arginine

Precaución: for Research Use Only

Formula: C₂₈H₃₀N₄O₆S
MW: 550.63 g/mol
MDL Number: MFCD00063331
CAS Number: 83792-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR