1-((2-(2,4-Difluorophenyl)oxiran-2-yl)methyl)-1H-1,2,4-triazole methanesulfonate 97%

Supplier: Ambeed
Warning

A151657-1G A151657-25G A151657-5G
77247-302EA 13 USD
77247-302 77247-304 77247-306
1-((2-(2,4-Difluorophenyl)oxiran-2-yl)methyl)-1H-1,2,4-triazole methanesulfonate 97%
1-((2-(2,4-Difluorophenyl)oxiran-2-yl)methyl)-1H-1,2,4-triazole methanesulfonate

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₂N₃O₄S
MW: 333.31 g/mol
MDL Number: MFCD13184268
CAS Number: 86386-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR