Refametinib 98+%

Supplier: Ambeed
Warning

Synonyms: BAY 86-9766, (S)-N-(3,4-Difluoro-2-((2-fluoro-4-iodophenyl)amino)-6-methoxyphenyl)-1-(2,3-dihydroxypropyl)cyclopropane-1-sulphonamide, N-[3,4-Difluoro-2-(2-fluoro-4-iodoanilino)-6-methoxyphenyl]-1-[(2S)-2,3-dihydroxypropyl]cyclopropane-1-sulphonamide

A151521-10MG A151521-1MG A151521-50MG A151521-5MG A151521-100MG
77247-286EA 54.59 USD
77247-286 77247-288 77247-290 77247-292 77746-046
Refametinib 98+%
Refametinib

Precaución: for Research Use Only

Formula: C₁₉H₂₀F₃IN₂O₅S
MW: 572.34 g/mol
MDL Number: MFCD18633256
CAS Number: 923032-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR