2-Fluoro-4-pyridone 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoropyridin-4-ol, 2-Fluoro-4-hydroxypyridine

A151419-100MG A151419-10G A151419-1G A151419-250MG A151419-5G
77259-258EA 15 USD
77259-258 77259-260 77259-262 77259-264 77259-266
2-Fluoro-4-pyridone 97%
2-Fluoro-4-pyridone

Precaución: for Research Use Only

Formula: C₅H₄FNO
MDL Number: MFCD04114159
CAS Number: 22282-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR