Ethyl 3-(3,4-dichlorophenyl)-3-oxopropanoate 97%

Supplier: Ambeed
Warning

A151366-100MG A151366-250MG A151366-1G
77272-998EA 26.78 USD
77272-998 77273-000 77748-046
Ethyl 3-(3,4-dichlorophenyl)-3-oxopropanoate 97%
Ethyl 3-(3,4-dichlorophenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₀Cl₂O₃
MW: 261.10 g/mol
MDL Number: MFCD03424830
CAS Number: 53090-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR