(R)-1-(5-Chloro-2-(2-(4-(4-fluorobenzyl)-2-methylpiperazin-1-yl)-2-oxoethoxy)phenyl)urea hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: BX471 HCl

A151361-10MG A151361-1MG A151361-25MG A151361-50MG A151361-5MG
77235-438EA 51.5 USD
77235-438 77235-440 77235-442 77235-444 77235-446
(R)-1-(5-Chloro-2-(2-(4-(4-fluorobenzyl)-2-methylpiperazin-1-yl)-2-oxoethoxy)phenyl)urea hydrochloride 98%
(R)-1-(5-Chloro-2-(2-(4-(4-fluorobenzyl)-2-methylpiperazin-1-yl)-2-oxoethoxy)phenyl)urea hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₅Cl₂FN₄O₃
MW: 471.35 g/mol
MDL Number: MFCD28167805
CAS Number: 288262-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR