3-(Ethoxycarbonyl)-4-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Ethoxycarbonyl-4-fluorophenylboronic acid, (3-Ethoxycarbonyl-4-fluorophenyl)boronic acid

A151360-1G A151360-250MG A151360-5G
77247-274EA 14.42 USD
77247-274 77247-276 77247-278
3-(Ethoxycarbonyl)-4-fluorophenylboronic acid 97%
3-(Ethoxycarbonyl)-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BFO₄
MW: 211.9827 g/mol
MDL Number: MFCD08061836
CAS Number: 874219-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR