Bilirubin 97%

Supplier: Ambeed
Warning

Synonyms: 3-(2-((3-(2-Carboxyethyl)-4-methyl-5-((Z)-(3-methyl-5-oxo-4-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-2-yl)methyl)-4-methyl-5-((Z)-(4-methyl-5-oxo-3-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-3-yl)propanoic acid

A151300-100MG A151300-1MG A151300-250MG
77286-112EA 33 USD
77286-112 77286-114 77286-116
Bilirubin 97%
Bilirubin

Precaución: for Research Use Only

Formula: C₃₃H₃₆N₄O₆
MW: 584.67 g/mol
MDL Number: MFCD00005499
CAS Number: 635-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR