2-Chloro-3-nitropyridine 98%

Supplier: Ambeed
Warning

A151249-1000G A151249-100G A151249-10G A151249-500G A151249-5G A151249-25G
77272-976EA 377 USD
77272-976 77272-978 77272-980 77272-982 77272-984 77299-862
2-Chloro-3-nitropyridine 98%
2-Chloro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₂
MDL Number: MFCD00006232
CAS Number: 5470-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR