4-Methoxy-2,6-dimethylphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethyl-4-methoxyphenylboronic acid

A151059-1G A151059-250MG A151059-5G
77247-260EA 29.87 USD
77247-260 77247-262 77549-902
4-Methoxy-2,6-dimethylphenylboronic acid 97%
4-Methoxy-2,6-dimethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
MDL Number: MFCD03452755
CAS Number: 361543-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR