(R)-2,2'-Bis(diphenylphosphino)-4,4',6,6'-tetramethoxy)-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

A151028-100MG A151028-1G A151028-250MG A151028-5G
77247-252EA 49.44 USD
77247-252 77247-254 77247-256 77247-258
(R)-2,2'-Bis(diphenylphosphino)-4,4',6,6'-tetramethoxy)-1,1'-biphenyl 98%
(R)-2,2'-Bis(diphenylphosphino)-4,4',6,6'-tetramethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₄₀H₃₆O₄P₂
MW: 642.66 g/mol
MDL Number: MFCD19443622
CAS Number: 1365531-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR