Iron(III) meso-tetraphenylporphine-µ-oxo dimer 97%

Supplier: Ambeed
Warning

Synonyms: Iron(III) meso-tetraphenylporphine-mu-oxo dimer, Meso-5,10,15,20-Tetraphenyl-21H,23H-porphineironu-oxodimer

A150939-100MG A150939-1G A150939-250MG A150939-5G
77247-244EA 27.81 USD
77247-244 77247-246 77247-248 77247-250
Iron(III) meso-tetraphenylporphine-µ-oxo dimer 97%
Iron(III) meso-tetraphenylporphine-µ-oxo dimer

Precaución: for Research Use Only

Formula: C₈₈H₅₆Fe₂N₈O
MW: 1353.13 g/mol
MDL Number: MFCD00058898
CAS Number: 12582-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR