2-Amino-2-(4-(heptyloxy)-3-(trifluoromethyl)phenethyl)propane-1,3-diol hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Amiselimod HCl

A150848-10MG A150848-1MG A150848-25MG A150848-5MG
77259-236EA 100.94 USD
77259-236 77259-238 77259-240 77259-242
2-Amino-2-(4-(heptyloxy)-3-(trifluoromethyl)phenethyl)propane-1,3-diol hydrochloride 98%
2-Amino-2-(4-(heptyloxy)-3-(trifluoromethyl)phenethyl)propane-1,3-diol hydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₃₁ClF₃NO₃
MW: 413.9 g/mol
MDL Number: MFCD25372041
CAS Number: 942398-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR