2-Chloro-5-fluorophenol 98%

Supplier: Ambeed
Warning

A150778-1G A150778-25G A150778-100G
77286-108EA 6 USD
77286-108 77286-110 77748-045
2-Chloro-5-fluorophenol 98%
2-Chloro-5-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₄ClFO
MW: 146.55
Boiling Pt: 183
MDL Number: MFCD00042524
CAS Number: 3827-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR