Sodium ((4-aminophenyl)sulfonyl)(6-methoxypyrimidin-4-yl)amide 97%

Supplier: Ambeed
Warning

Synonyms: sulphamonomethoxine sodium

A150700-1G A150700-250MG
77247-236EA 18.54 USD
77247-236 77247-238
Sodium ((4-aminophenyl)sulfonyl)(6-methoxypyrimidin-4-yl)amide 97%
Sodium ((4-aminophenyl)sulfonyl)(6-methoxypyrimidin-4-yl)amide

Precaución: for Research Use Only

Formula: C₁₁H₁₁N₄NaO₃S
MW: 302.29 g/mol
MDL Number: MFCD01750263
CAS Number: 38006-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR