Boc-L-homoserine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(Boc-amino)-4-hydroxybutyric acid, Boc-L-Homoserine, (S)-2-(tert-Butoxycarbonylamino)-4-hydroxybutyric acid, N-Boc-L-homoserine, N-(tert-Butoxycarbonyl)-L-homoserine

A150615-10G A150615-1G A150615-25G A150615-5G A150615-100G
77259-232EA 32 USD
77259-232 77259-234 77567-170 77567-172 77615-218
Boc-L-homoserine 97%
Boc-L-homoserine

Precaución: For research use only.

Formula: C₉H₁₇NO₅
MW: 219.2350 g/mol
MDL Number: MFCD00057839
CAS Number: 41088-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR