(S)-2-(4-Chlorophenyl)pyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

A150599-100MG A150599-1G A150599-250MG
77247-230EA 16 USD
77247-230 77247-232 77247-234
(S)-2-(4-Chlorophenyl)pyrrolidine hydrochloride 97%
(S)-2-(4-Chlorophenyl)pyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃Cl₂N
MDL Number: MFCD08751466
CAS Number: 1228560-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR