3-Iodo-7-methyl-1H-indazole 95%

Supplier: Ambeed
Warning

A150309-100MG A150309-1G A150309-250MG
77272-966EA 40 USD
77272-966 77272-968 77272-970
3-Iodo-7-methyl-1H-indazole 95%
3-Iodo-7-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇IN₂
MDL Number: MFCD07781632
CAS Number: 847906-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR