Methyl-2,5-dihydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Gentisic acid methyl ester, Methyl 2,5-dihydroxybenzoate, 2,5-Dihydroxybenzoic acid methyl ester

A150156-100G A150156-10G A150156-1G A150156-25G A150156-500G A150156-5G
77247-208EA 59.74 USD
77247-208 77247-210 77247-212 77247-214 77247-216 77247-218
Methyl-2,5-dihydroxybenzoate 98%
Methyl-2,5-dihydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00016464
CAS Number: 2150-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR