2-Isopropyl-4-nitrophenol 95%

Supplier: Ambeed
Danger

A150143-10G A150143-250MG A150143-5G
77259-222EA 137 USD
77259-222 77259-224 77259-226
2-Isopropyl-4-nitrophenol 95%
2-Isopropyl-4-nitrophenol

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MDL Number: MFCD09749960
CAS Number: 60515-72-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR