1-Cyclohexyl-4-ethylbenzene 97%

Supplier: Ambeed
Warning

A150058-1G A150058-250MG
77247-198EA 43 USD
77247-198 77247-200
1-Cyclohexyl-4-ethylbenzene 97%
1-Cyclohexyl-4-ethylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₂₀
MDL Number: MFCD29036695
CAS Number: 4501-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR