(S)-1-((S)-2-((Methoxycarbonyl)amino)-3-methylbutanoyl)pyrrolidine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A149892-100G A149892-1G A149892-25G A149892-5G
77259-200EA 122.57 USD
77259-200 77259-202 77259-204 77259-206
(S)-1-((S)-2-((Methoxycarbonyl)amino)-3-methylbutanoyl)pyrrolidine-2-carboxylic acid 95%
(S)-1-((S)-2-((Methoxycarbonyl)amino)-3-methylbutanoyl)pyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₅
MW: 272.30 g/mol
MDL Number: MFCD27946414
CAS Number: 181827-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR