2-Bromodibenzothiophene 98%

Supplier: Ambeed
Warning

A149860-10G A149860-1G A149860-25G A149860-5G
77259-192EA 11 USD
77259-192 77259-194 77259-196 77259-198
2-Bromodibenzothiophene 98%
2-Bromodibenzothiophene

Precaución: for Research Use Only

Formula: C₁₂H₇BrS
MDL Number: MFCD00089285
CAS Number: 22439-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR