2-Amino-2-(3-bromophenyl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-DL-phenylglycine

A149833-1G A149833-250MG A149833-25G A149833-5G
77235-400EA 51 USD
77235-400 77235-402 77235-404 77235-406
2-Amino-2-(3-bromophenyl)acetic acid 95%
2-Amino-2-(3-bromophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.0600 g/mol
MDL Number: MFCD00665358
CAS Number: 79422-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR