1-Methylindole-5-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A149663-1G A149663-250MG A149663-25G A149663-5G
77235-382EA 30 USD
77235-382 77235-384 77235-386 77235-388
1-Methylindole-5-boronic acid pinacol ester 97%
1-Methylindole-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₀BNO₂
MDL Number: MFCD05663865
CAS Number: 837392-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR