2-Aminomethyl-4-fluorobenzeneboronic acid hydrochloride 98%

Supplier: Ambeed
Warning

A149623-100MG A149623-1G A149623-250MG
77259-172EA 44 USD
77259-172 77259-174 77259-176
2-Aminomethyl-4-fluorobenzeneboronic acid hydrochloride 98%
2-Aminomethyl-4-fluorobenzeneboronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉BFNO₂·HCl
MDL Number: MFCD04973742
CAS Number: 850568-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR