NVP-231 98+%

Supplier: Ambeed
Warning

Synonyms: N -[2-(Benzoylamino)-6-benzothiazolyl]tricyclo[3.3.1.13,7]decane-1-carboxamide, N-(2-Benzamidobenzo[d]thiazol-6-yl)adamantane-1-carboxamide

A149574-10MG A149574-1MG A149574-50MG A149574-5MG
77235-370EA 29 USD
77235-370 77235-372 77235-374 77235-376
NVP-231 98+%
NVP-231

Precaución: for Research Use Only

Formula: C₂₅H₂₅N₃O₂S
MDL Number: MFCD01988824
CAS Number: 362003-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR