1-Methylpiperazine dihydrochloride 98%

Supplier: Ambeed
Warning

A149552-1G A149552-25G A149552-5G
77235-364EA 9 USD
77235-364 77235-366 77235-368
1-Methylpiperazine dihydrochloride 98%
1-Methylpiperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₄Cl₂N₂
MDL Number: MFCD00012755
CAS Number: 34352-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR