2-Ethoxy-5-((4-ethylpiperazin-1-yl)sulfonyl)benzoic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A149472-1G
A149472-250MG
77235-360EA
35.02
USD
77235-360
77235-362
2-Ethoxy-5-((4-ethylpiperazin-1-yl)sulfonyl)benzoic acid hydrochloride 97%
2-Ethoxy-5-((4-ethylpiperazin-1-yl)sulfonyl)benzoic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₁₅H₂₃ClN₂O₅S MW: 378.87 g/mol |
CAS Number:
1998216-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...