4-Bromopyridazine-3,6-diol 95%

Supplier: Ambeed
Warning

A149434-1G A149434-250MG
77259-168EA 30.9 USD
77259-168 77259-170
4-Bromopyridazine-3,6-diol 95%
4-Bromopyridazine-3,6-diol

Precaución: for Research Use Only

Formula: C₄H₃BrN₂O₂
MW: 190.98 g/mol
MDL Number: MFCD01077935
CAS Number: 43068-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR