(S)-4-Chloro-3-hydroxybutyronitrile 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-4-Chloro-3-hydroxybutyronitrile

A149282-10G A149282-5G A149282-500G
77286-070EA 6 USD
77286-070 77286-072 77567-112
(S)-4-Chloro-3-hydroxybutyronitrile 98%
(S)-4-Chloro-3-hydroxybutyronitrile

Precaución: For research use only.

Formula: C₄H₆ClNO
MW: 119.5500 g/mol
MDL Number: MFCD00273363
CAS Number: 127913-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR