2-Amino-4-methyl-5-pyridinecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-4-methylnicotinic acid

A149268-1G A149268-250MG A149268-25G
77247-190EA 77.25 USD
77247-190 77247-192 77247-194
2-Amino-4-methyl-5-pyridinecarboxylic acid 95%
2-Amino-4-methyl-5-pyridinecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.151 g/mol
MDL Number: MFCD09835242
CAS Number: 179555-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR